siRNA Details
| Gene Symbol: TSPYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022117.3 | NP_071400.1 | 1 | 546 | ||
| XM_006724592.3 | XP_006724655.1 | 1 | 579 | |||
| XM_017029726.1 | XP_016885215.1 | 1 | 579 | |||
| XM_017029727.1 | XP_016885216.1 | 1 | 579 | |||
| GenBank mRNA | AF254794.1 | - | 1 | 285 | ||
| AF273046.1 | - | 1 | 536 | |||
| AK131220.1 | - | 1 | 539 | |||
| AK292462.1 | - | 1 | 539 | |||
| AL713652.1 | - | 1 | 525 | |||
| AY040871.1 | - | 1 | 535 | |||
| BC024270.1 | - | 1 | 526 | 2819684 | ||

TSPYL2
Genomic Map