siRNA Details
| Gene Symbol: PRSS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022119.3 | NP_071402.1 | 3, 4 | 353 | ||
| XM_005255473.3 | XP_005255530.1 | 4, 5 | 452 | |||
| GenBank mRNA | AB010779.1 | - | 3, 4 | 290 | ||
| AF321182.1 | - | 3, 4 | 304 | |||
| AK309624.1 | - | 3, 4 | 495 | |||
| AY358396.1 | - | 3, 4 | 339 | |||
| BC009726.1 | - | 3, 4 | 325 | 3899480 | ||
| BQ893064.1 | - | 361 | ||||

PRSS22
Genomic Map