siRNA Details
| Gene Symbol: OXCT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022120.1 | NP_071403.1 | 1 | 756 | ||
| GenBank mRNA | AB050193.2 | - | 1 | 682 | ||
| AK093089.1 | - | 1 | 730 | |||
| AK314586.1 | - | 1 | 663 | |||
| AY013700.1 | - | 1 | 756 | |||
| BC030015.1 | - | 1 | 675 | |||
| Gene Symbol: OXCT2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132962.1 | 1 | 753 | |||
| GenBank mRNA | BC072388.1 | - | 1 | 753 | 5588771 | |

OXCT2
Genomic Map