siRNA Details
| Gene Symbol: SDF2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022044.2 | NP_071327.2 | 3 | 741 | ||
| GenBank mRNA | AB043007.1 | - | 3 | 677 | ||
| AF277316.1 | - | 3 | 665 | |||
| AY359118.1 | - | 3 | 708 | |||
| BC132849.1 | - | 3 | 733 | 40146871 | ||
| BC132851.1 | - | 3 | 733 | 40146873 | ||
| CR456570.1 | - | 3 | 666 | |||

SDF2L1
Genomic Map