siRNA Details
| Gene Symbol: DEF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022047.3 | NP_071330.3 | 3 | 437 | ||
| GenBank mRNA | AJ276095.1 | - | 3 | 398 | ||
| AK093613.1 | - | 3 | 437 | |||
| AK314756.1 | - | 3 | 412 | |||
| AY241694.1 | - | 3 | 437 | |||
| BC017504.1 | - | 3 | 403 | |||

DEF6
Genomic Map