siRNA Details
| Gene Symbol: FAM204A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134672.1 | NP_001128144.1 | 2 | 185 | ||
| NM_022063.2 | NP_071346.1 | 3 | 386 | |||
| XM_005270024.1 | XP_005270081.1 | 4 | 563 | |||
| GenBank mRNA | AK023250.1 | - | 3 | 365 | ||
| AK026630.1 | - | 3 | 379 | |||
| BC023577.2 | - | 3 | 369 | 4765095 | ||
| BM837313.1 | - | 2 | 178 | |||
| CR457304.1 | - | 1 | 119 | |||
| DA168924.1 | - | 3 | 390 | |||

FAM204A
Genomic Map