siRNA Details
| Gene Symbol: NAP1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021963.3 | NP_068798.1 | 1 | 1541 | ||
| GenBank mRNA | AB027013.1 | - | 1 | 1515 | ||
| AF136178.1 | - | 1 | 1489 | |||
| AK291297.1 | - | 1 | 1499 | |||
| AK303676.1 | - | 2 | 1134 | |||
| AK316569.1 | - | 1 | 1541 | |||
| BC026325.1 | - | 1 | 1463 | 4798964 | ||
| CR457095.1 | - | 1 | 1159 | |||

NAP1L2
Genomic Map