siRNA Details
| Gene Symbol: ITM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021999.4 | NP_068839.1 | 6 | 1056 | ||
| GenBank mRNA | AF086918.1 | - | 6 | 948 | ||
| AF092128.1 | - | 6 | 1045 | |||
| AF136973.1 | - | 6 | 1045 | |||
| AF152462.1 | - | 6 | 1003 | |||
| AF246221.1 | - | 6 | 1013 | |||
| AK000503.1 | - | 6 | 988 | |||
| AK027411.1 | - | 6 | 987 | |||
| AK130048.1 | - | 6 | 992 | |||
| BC000554.2 | - | 6 | 982 | 3163436 | ||
| BC016148.1 | - | 6 | 1003 | 3912066 | ||

ITM2B
Genomic Map