siRNA Details
| Gene Symbol: DEF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022047.3 | NP_071330.3 | 11 | 1910 | ||
| GenBank mRNA | AJ276095.1 | - | 11 | 1871 | ||
| AK093613.1 | - | 10 | 1622 | |||
| AK314756.1 | - | 11 | 1885 | |||
| AL110182.1 | - | 1 | 112 | |||
| AY241694.1 | - | 11 | 1910 | |||
| BC017504.1 | - | 11 | 1876 | |||

DEF6
Genomic Map