siRNA Details
| Gene Symbol: THADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083953.1 | NP_001077422.1 | 38 | 6282 | ||
| NM_022065.4 | NP_071348.3 | 38 | 6070 | |||
| NR_073394.1 | 36 | 5880 | ||||
| XM_006712061.3 | XP_006712124.1 | 39 | 6117 | |||
| XM_006712062.1 | XP_006712125.1 | 38 | 6067 | |||
| XM_006712063.1 | XP_006712126.1 | 38 | 5950 | |||
| XM_006712065.1 | XP_006712128.1 | 36 | 5848 | |||
| XM_006712066.1 | XP_006712129.1 | 36 | 5707 | |||
| XM_006712067.1 | XP_006712130.1 | 36 | 5704 | |||
| XM_006712069.3 | XP_006712132.1 | 23 | 3578 | |||
| XM_017004674.1 | XP_016860163.1 | 38 | 5947 | |||
| XM_017004675.1 | XP_016860164.1 | 37 | 5822 | |||
| GenBank mRNA | AB051554.2 | - | 38 | 6282 | ||
| AK025445.1 | - | 23 | 3649 | |||
| AK025530.1 | - | 13 | 2143 | |||
| AK290023.1 | - | 38 | 6070 | |||
| AY149629.1 | - | 38 | 6062 | |||
| AY149633.1 | - | 36 | 5872 | |||
| BC007743.1 | - | 9 | 1664 | |||
| BC025773.1 | - | 14 | 2283 | |||
| BC105976.1 | - | 17 | 2580 | |||
| BX648155.1 | - | 38 | 6066 | |||

THADA
Genomic Map