siRNA Details
| Gene Symbol: SLC17A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302643.1 | NP_001289572.1 | 14 | 2574 | ||
| NM_022082.3 | NP_071365.3 | 13 | 2512 | |||
| XM_011528978.2 | XP_011527280.1 | 13 | 2424 | |||
| GenBank mRNA | AK027065.1 | - | 6 | 2047 | ||
| AK074107.1 | - | 11 | 3912 | |||
| AK095473.1 | - | 13 | 2363 | |||
| AK125597.1 | - | 13 | 3035 | |||
| BC025312.1 | - | 14 | 2514 | 4563089 | ||
| BC027447.1 | - | 9 | 1733 | |||
| BC038593.1 | - | 13 | 2354 | |||

SLC17A9
Genomic Map