siRNA Details
| Gene Symbol: ZNF335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022095.3 | NP_071378.1 | 6, 7 | 1085 | ||
| XM_005260504.4 | XP_005260561.1 | 6, 7 | 1798 | |||
| XM_011528979.2 | XP_011527281.1 | 5, 6 | 1453 | |||
| XM_017028012.1 | XP_016883501.1 | 5, 6 | 856 | |||
| GenBank mRNA | AF395833.1 | - | 6, 7 | 1062 | ||
| AK092485.1 | - | 4, 5 | 1294 | |||
| AK296989.1 | - | 3, 4 | 656 | |||
| BC041331.2 | - | 3, 4 | 300 | |||
| BC142981.1 | - | 3, 4 | 300 | |||

ZNF335
Genomic Map