siRNA Details
| Gene Symbol: CHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022097.3 | NP_071380.1 | 7 | 1780 | ||
| GenBank mRNA | AF146019.1 | - | 7 | 1785 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHP2| Gene Symbol: CHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022097.3 | NP_071380.1 | 7 | 1780 | ||
| GenBank mRNA | AF146019.1 | - | 7 | 1785 | ||