siRNA Details
| Gene Symbol: PRSS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022119.3 | NP_071402.1 | 5 | 671 | ||
| XM_005255473.3 | XP_005255530.1 | 6 | 770 | |||
| GenBank mRNA | AB010779.1 | - | 5 | 608 | ||
| AF321182.1 | - | 5 | 622 | |||
| AK309624.1 | - | 5 | 813 | |||
| AY358396.1 | - | 5 | 657 | |||
| BC009726.1 | - | 5 | 643 | 3899480 | ||

PRSS22
Genomic Map