siRNA Details
| Gene Symbol: LRRC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022143.4 | NP_071426.1 | 2 | 2393 | ||
| XM_011516461.2 | XP_011514763.1 | 3 | 2699 | |||
| XM_017012505.1 | XP_016867994.1 | 3 | 2589 | |||
| GenBank mRNA | AF196976.4 | - | 2 | 2396 | ||
| AK172751.1 | - | 2 | 2397 | |||
| BC036685.1 | - | 1 | 67 | |||
| BC111561.1 | - | 2 | 2387 | 40068142 | ||
| BC111745.1 | - | 2 | 2387 | 40068146 | ||

LRRC4
Genomic Map