siRNA Details
| Gene Symbol: ROBO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022370.3 | NP_071765.2 | 27, 28 | 4326 | ||
| XM_017018122.1 | XP_016873611.1 | 26, 27 | 4091 | |||
| GenBank mRNA | AK024697.1 | - | 11, 12 | 1557 | ||
| AK225076.1 | - | 11, 12 | 1557 | |||
| AK298792.1 | - | 10, 11 | 1426 | |||
| AY509035.1 | - | 27, 28 | 4349 | |||
| BC008623.1 | - | 7, 8 | 1297 | |||

ROBO3
Genomic Map