siRNA Details
| Gene Symbol: OCLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022375.3 | NP_071770.1 | 1 | 873 | ||
| GenBank mRNA | AF142063.1 | - | 1 | 988 | ||
| BC069096.1 | - | 1 | 890 | 7216910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OCLM| Gene Symbol: OCLM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022375.3 | NP_071770.1 | 1 | 873 | ||
| GenBank mRNA | AF142063.1 | - | 1 | 988 | ||
| BC069096.1 | - | 1 | 890 | 7216910 | ||