siRNA Details
| Gene Symbol: LMBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022458.3 | NP_071903.2 | 3, 4 | 396 | ||
| XM_005249555.2 | XP_005249612.1 | 3, 4 | 371 | |||
| XM_005249558.2 | XP_005249615.1 | 3, 4 | 371 | |||
| XM_017012507.1 | XP_016867996.1 | 3, 4 | 371 | |||
| XM_017012509.1 | XP_016867998.1 | 3, 4 | 371 | |||
| XM_017012514.1 | XP_016868003.1 | 4, 5 | 465 | |||
| XM_017012515.1 | XP_016868004.1 | 3, 4 | 371 | |||
| XM_017012517.1 | XP_016868006.1 | 3, 4 | 371 | |||
| XM_017012518.1 | XP_016868007.1 | 4, 5 | 469 | |||
| GenBank mRNA | AF402318.1 | - | 3, 4 | 207 | ||
| AK021727.1 | - | 3, 4 | 367 | |||
| AK291037.1 | - | 3, 4 | 367 | |||
| AK299753.1 | - | 2, 3 | 294 | |||
| AL834394.1 | - | 3, 4 | 175 | |||
| BC017663.1 | - | 3, 4 | 335 | 3883022 | ||
| BC063649.1 | - | 4, 5 | 465 | |||
| DA238712.1 | - | 3, 4 | 396 | |||

LMBR1
Genomic Map