siRNA Details
| Gene Symbol: TCTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022171.2 | NP_071503.1 | 3 | 1253 | ||
| GenBank mRNA | AK000824.1 | - | 3 | 1070 | ||
| EL954558.1 | - | 1 | 242 | |||
| L41143.1 | - | 3 | 1253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCTA| Gene Symbol: TCTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022171.2 | NP_071503.1 | 3 | 1253 | ||
| GenBank mRNA | AK000824.1 | - | 3 | 1070 | ||
| EL954558.1 | - | 1 | 242 | |||
| L41143.1 | - | 3 | 1253 | |||