siRNA Details
| Gene Symbol: RAB38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022337.2 | NP_071732.1 | 2 | 321 | ||
| XM_017017455.1 | XP_016872944.1 | 2 | 359 | |||
| XM_017017456.1 | XP_016872945.1 | 2 | 359 | |||
| GenBank mRNA | AF136172.1 | - | 2 | 242 | ||
| AF235022.1 | - | 2 | 285 | |||
| AL546159.3 | - | 2 | 321 | |||
| BC015808.1 | - | 2 | 287 | 3916084 | ||
| BT009842.1 | - | 2 | 238 | |||

RAB38
Genomic Map