siRNA Details
| Gene Symbol: SPATA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310156.1 | NP_001297085.1 | 5 | 530 | ||
| XM_011542514.2 | XP_011540816.1 | 5 | 530 | |||
| XM_011542515.2 | XP_011540817.1 | 5 | 530 | |||
| XM_011542516.1 | XP_011540818.1 | 5 | 530 | |||
| XM_011542517.1 | XP_011540819.1 | 5 | 530 | |||
| XM_011542519.2 | XP_011540821.1 | 5 | 530 | |||
| XM_011542520.2 | XP_011540822.1 | 5 | 530 | |||
| XM_011542521.2 | XP_011540823.1 | 5 | 530 | |||
| XM_011542522.2 | XP_011540824.1 | 5 | 530 | |||
| GenBank mRNA | AF306347.1 | - | 5 | 502 | ||
| BC119641.1 | - | 5 | 434 | |||
| BC119642.1 | - | 5 | 434 | |||
| BC127634.1 | - | 5 | 434 | 40115717 | ||
| CD300599.1 | - | 5 | 546 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027617, BC028997, BC055056, BC042647, BC139279, and others.

SPATA1
Genomic Map