siRNA Details
| Gene Symbol: POPDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022361.4 | NP_071756.2 | 2 | 489 | ||
| NR_024539.1 | 2 | 401 | ||||
| XM_011536067.2 | XP_011534369.1 | 2 | 649 | |||
| XM_017011194.1 | XP_016866683.1 | 2 | 1932 | |||
| XM_017011195.1 | XP_016866684.1 | 2 | 1392 | |||
| GenBank mRNA | AF204171.1 | - | 1 | 234 | ||
| AK225989.1 | - | 2 | 401 | |||
| AK314101.1 | - | 2 | 489 | |||
| BC022323.2 | - | 1 | 339 | 4293961 | ||
| BF244570.1 | - | 501 | ||||

POPDC3
Genomic Map