siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 18 | 3037 | ||
| XM_011545916.2 | XP_011544218.1 | 17 | 4384 | |||
| XM_011545917.2 | XP_011544219.1 | 17 | 4384 | |||
| XM_011545919.2 | XP_011544221.1 | 16 | 4347 | |||
| GenBank mRNA | AK022551.1 | - | 9 | 1442 | ||
| AK093045.1 | - | 2 | 1713 | |||
| AK096322.1 | - | 10 | 1820 | |||
| AK298229.1 | - | 11 | 1600 | |||

FBRS
Genomic Map