siRNA Details
| Gene Symbol: ZNF106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022473.2 | NP_071918.1 | 1 | 556 | ||
| XM_005254591.2 | XP_005254648.1 | 4 | 463 | |||
| XM_005254592.2 | XP_005254649.1 | 4 | 463 | |||
| GenBank mRNA | AF205632.1 | - | 1 | 529 | ||
| AK091929.1 | - | 2 | 292 | |||
| BC029366.1 | - | 1 | 520 | |||
| BC058023.1 | - | 1 | 525 | |||
| BC070244.1 | - | 1 | 520 | |||
| BC111396.1 | - | 1 | 525 | |||
| BX649141.1 | - | 4 | 440 | |||

ZNF106
Genomic Map