siRNA Details
Gene Symbol: RTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022147.2 | NP_071430.2 | 2 | 911 | ||
GenBank mRNA | AJ251832.1 | - | 2 | 903 | ||
BC013161.1 | - | 2 | 911 | 3919961 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022147.2 | NP_071430.2 | 2 | 911 | ||
GenBank mRNA | AJ251832.1 | - | 2 | 903 | ||
BC013161.1 | - | 2 | 911 | 3919961 |