siRNA Details
| Gene Symbol: RRAGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271851.1 | NP_001258780.1 | 6 | 1070 | ||
| NM_022157.3 | NP_071440.1 | 6 | 1172 | |||
| GenBank mRNA | AF272035.1 | - | 6 | 1000 | ||
| AF323609.1 | - | 6 | 995 | |||
| AK023373.1 | - | 2 | 200 | |||
| AK092808.1 | - | 1 | 2083 | |||
| AK298573.1 | - | 5 | 695 | |||
| AK298788.1 | - | 6 | 966 | |||
| AK315791.1 | - | 6 | 1096 | |||
| BC016668.1 | - | 6 | 1017 | 4342448 | ||
| CX761032.1 | - | 1 | 82 | |||

RRAGC
Genomic Map