siRNA Details
| Gene Symbol: RBSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302378.1 | NP_001289307.1 | 4 | 637 | ||
| NM_022340.3 | NP_071735.2 | 5 | 793 | |||
| XM_005265384.4 | XP_005265441.1 | 5 | 756 | |||
| XM_005265385.4 | XP_005265442.1 | 4 | 611 | |||
| XM_017007023.1 | XP_016862512.1 | 4 | 629 | |||
| XM_017007024.1 | XP_016862513.1 | 6 | 939 | |||
| XM_017007025.1 | XP_016862514.1 | 5 | 798 | |||
| XM_017007026.1 | XP_016862515.1 | 4 | 643 | |||
| XM_017007027.1 | XP_016862516.1 | 5 | 784 | |||
| XM_017007028.1 | XP_016862517.1 | 5 | 784 | |||
| GenBank mRNA | AB209673.1 | - | 5 | 753 | ||
| AK291288.1 | - | 4 | 628 | |||
| AK301735.1 | - | 4 | 629 | |||
| AY009133.1 | - | 2 | 170 | |||
| BC106940.2 | - | 2 | 170 | 40034008 | ||

RBSN
Genomic Map