siRNA Details
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022473.2 | NP_071918.1 | 2 | 2404 | ||
XM_005254591.2 | XP_005254648.1 | 5 | 2311 | |||
XM_005254592.2 | XP_005254649.1 | 5 | 2311 | |||
XM_017022483.1 | XP_016877972.1 | 4 | 2086 | |||
XM_017022484.1 | XP_016877973.1 | 2 | 1831 | |||
GenBank mRNA | AF205632.1 | - | 2 | 2377 | ||
AK057410.1 | - | 1 | 1385 | |||
AK091929.1 | - | 3 | 2140 | |||
AK294529.1 | - | 2 | 2134 | |||
AL832352.1 | - | 1 | 404 | |||
AL832455.2 | - | 1 | 318 | |||
AL833301.1 | - | 1 | 415 | |||
BX648629.1 | - | 1 | 1509 |
