siRNA Details
| Gene Symbol: ACBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022735.3 | NP_073572.2 | 2 | 448 | ||
| GenBank mRNA | AB043587.2 | - | 2 | 456 | ||
| AK025520.1 | - | 2 | 449 | |||
| AK314468.1 | - | 2 | 449 | |||
| AY150218.1 | - | 2 | 427 | |||
| BC045533.1 | - | 2 | 434 | 5259930 | ||
| BC060792.1 | - | 2 | 444 | 4813993 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_133225, AK030371, AK168799, BC050121, AK169505, and others.

ACBD3
Genomic Map