siRNA Details
| Gene Symbol: IFT22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130821.2 | NP_001124293.1 | 2, 3 | 262 | ||
| NM_001287525.1 | NP_001274454.1 | 3, 4 | 304 | |||
| NM_022777.3 | NP_073614.1 | 2, 3 | 231 | |||
| GenBank mRNA | AK023287.1 | - | 3, 4 | 262 | ||
| AK024179.1 | - | 2, 3 | 157 | |||
| AK294653.1 | - | 2, 3 | 216 | |||
| AU133515.1 | - | 3, 4 | 262 | |||
| BC009823.2 | - | 2, 3 | 204 | 3946565 | ||
| BU597007.1 | - | 2, 3 | 239 | |||
| CA396999.1 | - | 2, 3 | 220 | |||
| DN993868.1 | - | 2, 3 | 240 | |||

IFT22
Genomic Map