siRNA Details
| Gene Symbol: GIGYF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022574.4 | NP_072096.2 | 8 | 1914 | ||
| XM_005250532.1 | XP_005250589.1 | 10 | 1695 | |||
| XM_011516472.1 | XP_011514774.1 | 10 | 1695 | |||
| XM_011516477.1 | XP_011514779.1 | 10 | 1695 | |||
| XM_017012526.1 | XP_016868015.1 | 10 | 1695 | |||
| XM_017012527.1 | XP_016868016.1 | 12 | 1593 | |||
| XM_017012528.1 | XP_016868017.1 | 11 | 1205 | |||
| XM_017012529.1 | XP_016868018.1 | 11 | 1212 | |||
| XM_017012530.1 | XP_016868019.1 | 11 | 1466 | |||
| XM_017012531.1 | XP_016868020.1 | 6 | 718 | |||
| GenBank mRNA | AY176044.1 | - | 8 | 2115 | ||
| BC129991.1 | - | 2 | 62 | |||

GIGYF1
Genomic Map