siRNA Details
Gene Symbol: FANCF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022725.3 | NP_073562.1 | 1 | 516 | ||
GenBank mRNA | AF181994.1 | - | 1 | 489 | ||
AF181995.1 | - | 1 | 485 | |||
AK001716.2 | - | 1 | 498 | |||
AK023153.1 | - | 1 | 498 | |||
AK223277.1 | - | 1 | 490 | |||
BC035346.1 | - | 1 | 510 | |||
BC047028.1 | - | 1 | 488 | 5538578 | ||
BC063038.1 | - | 1 | 516 | 5239725 | ||
BC093867.1 | - | 1 | 506 | 7939712 | ||
BC101807.1 | - | 1 | 506 | 8069313 | ||
BI522448.1 | - | 1 | 516 | |||
BX640996.1 | - | 1 | 508 |
