siRNA Details
| Gene Symbol: MARC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022746.3 | NP_073583.3 | 4 | 969 | ||
| XM_011509900.2 | XP_011508202.1 | 4 | 969 | |||
| XM_011509903.2 | XP_011508205.1 | 4 | 1345 | |||
| XM_011509904.2 | XP_011508206.1 | 4 | 622 | |||
| XM_017002096.1 | XP_016857585.1 | 4 | 549 | |||
| XM_017002097.1 | XP_016857586.1 | 4 | 602 | |||
| GenBank mRNA | AK026043.1 | - | 4 | 765 | ||
| AK092439.1 | - | 3 | 1440 | |||
| AK094105.1 | - | 5 | 632 | |||
| AK225416.1 | - | 4 | 765 | |||
| AK290812.1 | - | 4 | 741 | |||
| BC010619.1 | - | 4 | 737 | 3872779 | ||

MARC1
Genomic Map