siRNA Details
| Gene Symbol: C6orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022758.5 | NP_073595.2 | 4 | 3412 | ||
| NM_024294.3 | NP_077270.1 | 5 | 3610 | |||
| GenBank mRNA | AF370387.1 | - | 1 | 356 | ||
| AK025848.1 | - | 1 | 663 | |||
| BC010143.2 | - | 1 | 846 | |||
| BC013241.1 | - | 1 | 1047 | |||
| BC075810.1 | - | 5 | 3532 | 5535440 | ||
| BC107732.1 | - | 1 | 492 | |||

C6orf106
Genomic Map