siRNA Details
| Gene Symbol: OSBPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022776.4 | NP_073613.2 | 1, 2 | 896 | ||
| GenBank mRNA | AF346292.1 | - | 1, 2 | 323 | ||
| AF392454.1 | - | 1, 2 | 467 | |||
| AK292702.1 | - | 1, 2 | 248 | |||
| BC065213.1 | - | 1, 2 | 246 | 3916115 | ||
| BX506747.1 | - | 1, 2 | 672 | |||

OSBPL11
Genomic Map