siRNA Details
Gene Symbol: RNF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022781.4 | NP_073618.3 | 4 | 585 | ||
NM_194328.2 | NP_919309.1 | 4 | 752 | |||
NM_194329.2 | NP_919310.1 | 3 | 435 | |||
NM_194330.2 | NP_919311.1 | 4 | 483 | |||
NM_194332.2 | NP_919313.1 | 3 | 602 | |||
XM_005251366.3 | XP_005251423.1 | 4 | 589 | |||
XM_005251367.3 | XP_005251424.1 | 4 | 487 | |||
XM_005251368.3 | XP_005251425.1 | 4 | 601 | |||
XM_006716721.3 | XP_006716784.1 | 3 | 449 | |||
XM_011517712.2 | XP_011516014.1 | 5 | 525 | |||
XM_011517713.2 | XP_011516015.1 | 4 | 589 | |||
XM_017014294.1 | XP_016869783.1 | 3 | 506 | |||
XM_017014295.1 | XP_016869784.1 | 4 | 616 | |||
XM_017014296.1 | XP_016869785.1 | 3 | 186 | |||
XM_017014297.1 | XP_016869786.1 | 3 | 182 | |||
GenBank mRNA | AF394047.1 | - | 4 | 707 | ||
AK024996.1 | - | 2 | 95 | |||
AK093480.1 | - | 4 | 651 | |||
AK097653.1 | - | 4 | 645 | |||
AL136817.1 | - | 4 | 503 | |||
AV690059.2 | - | 3 | 326 | |||
AX748155.1 | - | 4 | 651 | |||
BC033786.1 | - | 3 | 435 | 5166325 | ||
BG722531.1 | - | 3 | 608 | |||
BI459742.1 | - | 3 | 622 | |||
BI520161.1 | - | 4 | 481 | |||
DA189510.1 | - | 3 | 553 | |||
DA483270.1 | - | 3 | 333 |
