siRNA Details
| Gene Symbol: GIGYF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022574.4 | NP_072096.2 | 24 | 4445 | ||
| XM_005250532.1 | XP_005250589.1 | 26 | 4226 | |||
| XM_011516472.1 | XP_011514774.1 | 26 | 4243 | |||
| XM_011516477.1 | XP_011514779.1 | 26 | 4240 | |||
| XM_017012526.1 | XP_016868015.1 | 25 | 4318 | |||
| XM_017012527.1 | XP_016868016.1 | 27 | 4216 | |||
| XM_017012528.1 | XP_016868017.1 | 26 | 3828 | |||
| XM_017012529.1 | XP_016868018.1 | 26 | 3835 | |||
| XM_017012530.1 | XP_016868019.1 | 26 | 4089 | |||
| XM_017012531.1 | XP_016868020.1 | 21 | 3341 | |||
| GenBank mRNA | AF086157.1 | - | 2 | 419 | ||
| AF289554.1 | - | 6 | 1250 | |||
| AY176044.1 | - | 24 | 4646 | |||
| BC018563.1 | - | 8 | 1418 | |||
| BC036595.2 | - | 1 | 290 | |||

GIGYF1
Genomic Map