siRNA Details
| Gene Symbol: FANCF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022725.3 | NP_073562.1 | 1 | 1579 | ||
| GenBank mRNA | AK001716.2 | - | 1 | 1561 | ||
| AK023153.1 | - | 1 | 1561 | |||
| AK223277.1 | - | 1 | 1553 | |||
| BC047028.1 | - | 1 | 1551 | 5538578 | ||
| BC063038.1 | - | 1 | 1579 | 5239725 | ||
| BX640996.1 | - | 1 | 1571 | |||

FANCF
Genomic Map