siRNA Details
Gene Symbol: RNF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022781.4 | NP_073618.3 | 12 | 4546 | ||
NM_194328.2 | NP_919309.1 | 12 | 4713 | |||
NM_194329.2 | NP_919310.1 | 11 | 4396 | |||
NM_194330.2 | NP_919311.1 | 12 | 4444 | |||
NM_194332.2 | NP_919313.1 | 11 | 4563 | |||
XM_005251366.3 | XP_005251423.1 | 12 | 4550 | |||
XM_005251367.3 | XP_005251424.1 | 12 | 4448 | |||
XM_005251368.3 | XP_005251425.1 | 12 | 4562 | |||
XM_006716721.3 | XP_006716784.1 | 11 | 4410 | |||
XM_011517712.2 | XP_011516014.1 | 13 | 4486 | |||
XM_011517713.2 | XP_011516015.1 | 12 | 4550 | |||
XM_017014294.1 | XP_016869783.1 | 11 | 4467 | |||
XM_017014295.1 | XP_016869784.1 | 12 | 4577 | |||
XM_017014296.1 | XP_016869785.1 | 11 | 4147 | |||
XM_017014297.1 | XP_016869786.1 | 11 | 4143 | |||
GenBank mRNA | AF394047.1 | - | 13 | 4136 | ||
AK024996.1 | - | 11 | 3524 | |||
AK025466.1 | - | 1 | 2574 |
