siRNA Details
| Gene Symbol: MARCH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282805.1 | NP_001269734.1 | 12 | 2764 | ||
| NM_001282806.1 | NP_001269735.1 | 8 | 2419 | |||
| NM_001282807.1 | NP_001269736.1 | 9 | 2511 | |||
| NM_022826.3 | NP_073737.1 | 10 | 2636 | |||
| XM_005246773.2 | XP_005246830.1 | 10 | 2659 | |||
| XM_005246774.1 | XP_005246831.1 | 9 | 2610 | |||
| XM_017004718.1 | XP_016860207.1 | 9 | 2587 | |||
| XM_017004719.1 | XP_016860208.1 | 9 | 2494 | |||
| XM_017004720.1 | XP_016860209.1 | 9 | 2456 | |||
| XM_017004721.1 | XP_016860210.1 | 8 | 2437 | |||
| GenBank mRNA | AK022973.1 | - | 10 | 2594 | ||
| AK124225.1 | - | 1 | 3117 | |||
| AL050171.1 | - | 3 | 598 | |||
| AL833651.1 | - | 6 | 1660 | |||
| BC003404.1 | - | 6 | 1035 | |||
| BC065014.1 | - | 10 | 2560 | 6169288 | ||
| CR627026.1 | - | 1 | 2736 | |||

MARCH7
Genomic Map