siRNA Details
| Gene Symbol: SLC13A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011554.2 | NP_001011554.1 | 8 | 1013 | ||
| NM_001193339.1 | NP_001180268.1 | 6 | 827 | |||
| NM_001193340.1 | NP_001180269.1 | 8 | 1013 | |||
| NM_001193342.1 | NP_001180271.1 | 7 | 804 | |||
| NM_022829.5 | NP_073740.2 | 7 | 977 | |||
| GenBank mRNA | AF154121.1 | - | 7 | 956 | ||
| AK056713.1 | - | 7 | 975 | |||
| AK096658.1 | - | 8 | 1013 | |||
| AK293902.1 | - | 7 | 793 | |||
| AK295748.1 | - | 6 | 825 | |||
| AK310960.1 | - | 6 | 987 | |||
| AK315131.1 | - | 7 | 975 | |||
| AY072810.3 | - | 7 | 956 | |||
| BC035966.1 | - | 7 | 975 | 4609776 | ||
| BM474324.1 | - | 408 | ||||
| DA272524.1 | - | 2 | 154 | |||

SLC13A3
Genomic Map