siRNA Details
| Gene Symbol: TMEM135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168724.1 | NP_001162195.1 | 5, 6 | 653 | ||
| NM_022918.3 | NP_075069.3 | 6, 7 | 719 | |||
| NR_033149.1 | 5, 6 | 685 | ||||
| XM_017018140.1 | XP_016873629.1 | 5, 6 | 584 | |||
| XM_017018141.1 | XP_016873630.1 | 4, 5 | 268 | |||
| XM_017018142.1 | XP_016873631.1 | 4, 5 | 221 | |||
| GenBank mRNA | AK000684.1 | - | 3, 4 | 263 | ||
| AK025757.1 | - | 4, 5 | 267 | |||
| AK225364.1 | - | 4, 5 | 267 | |||
| AK291804.1 | - | 5, 6 | 563 | |||
| AK296774.1 | - | 2, 3 | 257 | |||
| AK302473.1 | - | 5, 6 | 489 | |||
| BC051462.1 | - | 6, 7 | 540 | 5278478 | ||
| BX648678.1 | - | 5, 6 | 639 | |||

TMEM135
Genomic Map