siRNA Details
| Gene Symbol: DDRGK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023935.2 | NP_076424.1 | 2 | 354 | ||
| GenBank mRNA | AK310809.1 | - | 2 | 337 | ||
| BC000643.1 | - | 2 | 306 | 3344932 | ||
| BC007957.1 | - | 2 | 312 | 4301923 | ||
| BC011851.1 | - | 2 | 307 | 4136696 | ||

DDRGK1
Genomic Map