siRNA Details
| Gene Symbol: NKX3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256339.1 | NP_001243268.1 | 3 | 1413 | ||
| NM_006167.3 | NP_006158.2 | 2 | 1686 | |||
| NR_046072.1 | 1 | 873 | ||||
| GenBank mRNA | AF247704.1 | - | 2 | 1685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKX3-1| Gene Symbol: NKX3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256339.1 | NP_001243268.1 | 3 | 1413 | ||
| NM_006167.3 | NP_006158.2 | 2 | 1686 | |||
| NR_046072.1 | 1 | 873 | ||||
| GenBank mRNA | AF247704.1 | - | 2 | 1685 | ||