siRNA Details
| Gene Symbol: YIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321439.1 | NP_001308368.1 | 7 | 691 | ||
| NM_001321440.1 | NP_001308369.1 | 6 | 574 | |||
| NM_024029.4 | NP_076934.1 | 7 | 962 | |||
| XM_011528270.2 | XP_011526572.1 | 7 | 691 | |||
| XM_011528271.2 | XP_011526573.1 | 6 | 552 | |||
| GenBank mRNA | BC000056.2 | - | 7 | 610 | 3506385 | |
| BC013014.2 | - | 7 | 631 | 2960233 | ||
| BM554546.1 | - | 6 | 492 | |||

YIPF2
Genomic Map