siRNA Details
| Gene Symbol: AUNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287490.1 | NP_001274419.1 | 2 | 206 | ||
| NM_024037.2 | NP_076942.1 | 2 | 206 | |||
| XM_017002300.1 | XP_016857789.1 | 2 | 277 | |||
| GenBank mRNA | AK024326.1 | - | 2 | 105 | ||
| AK223419.1 | - | 2 | 142 | |||
| BC000209.1 | - | 2 | 160 | 3350471 | ||
| BJ989876.1 | - | 2 | 215 | |||
| GQ844687.1 | - | 2 | 160 | |||

AUNIP
Genomic Map