siRNA Details
| Gene Symbol: GGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199815.1 | NP_001186744.1 | 1, 2 | 281 | ||
| NM_001199816.1 | NP_001186745.1 | 1, 2 | 281 | |||
| NM_024051.3 | NP_076956.1 | 1, 2 | 281 | |||
| GenBank mRNA | AK315608.1 | - | 1, 2 | 272 | ||
| BC000625.2 | - | 1, 2 | 276 | 3346822 | ||
| BC005356.1 | - | 1, 2 | 269 | 4102029 | ||
| BC013937.1 | - | 1, 2 | 242 | 3997629 | ||
| BC019243.1 | - | 1, 2 | 273 | 3828006 | ||
| BE407390.1 | - | 1, 2 | 210 | |||

GGCT
Genomic Map