siRNA Details
| Gene Symbol: TMEM135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168724.1 | NP_001162195.1 | 6 | 660 | ||
| NM_022918.3 | NP_075069.3 | 7 | 726 | |||
| NR_033149.1 | 6 | 692 | ||||
| XM_017018140.1 | XP_016873629.1 | 6 | 591 | |||
| XM_017018141.1 | XP_016873630.1 | 5 | 275 | |||
| XM_017018142.1 | XP_016873631.1 | 5 | 228 | |||
| GenBank mRNA | AK000684.1 | - | 4 | 270 | ||
| AK025757.1 | - | 5 | 274 | |||
| AK225364.1 | - | 5 | 274 | |||
| AK291804.1 | - | 6 | 570 | |||
| AK296774.1 | - | 3 | 264 | |||
| AK302473.1 | - | 6 | 496 | |||
| BC051462.1 | - | 7 | 547 | 5278478 | ||
| BX648678.1 | - | 6 | 646 | |||

TMEM135
Genomic Map