siRNA Details
| Gene Symbol: DNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172810.1 | NP_001166281.1 | 4 | 550 | ||
| NM_023036.4 | NP_075462.3 | 4 | 553 | |||
| XM_011525125.2 | XP_011523427.1 | 4 | 570 | |||
| GenBank mRNA | AF250288.1 | - | 4 | 510 | ||
| AJ295276.1 | - | 3 | 749 | |||
| BC039582.1 | - | 4 | 540 | 5752153 | ||
| BI464166.1 | - | 4 | 539 | |||

DNAI2
Genomic Map