siRNA Details
| Gene Symbol: UBE2Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023079.4 | NP_075567.2 | 5 | 846 | ||
| GenBank mRNA | AB025426.1 | - | 4 | 387 | ||
| AK023917.1 | - | 4 | 384 | |||
| AK024030.1 | - | 6 | 701 | |||
| AK091644.1 | - | 3 | 1977 | |||
| AK122770.1 | - | 5 | 744 | |||
| AK130303.1 | - | 6 | 704 | |||
| BC015169.1 | - | 4 | 373 | |||
| BC015890.1 | - | 5 | 714 | 3915698 | ||
| CR457322.1 | - | 4 | 387 | |||
| EF623992.1 | - | 5 | 711 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172300, AK005524, AK152529, BC027266, AK153328, and others.

UBE2Z
Genomic Map